2-Ethoxybenzoic acid ≥98%

147494-100G 147494-10G
220037-108EA 90.78 USD
220037-108 220037-109
2-Ethoxybenzoic acid ≥98%
2-Ethoxybenzoic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.173 g/cm³ (20 °C)
MDL Number: MFCD00002438
CAS Number: 134-11-2
EINECS: 205-130-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR