Benzyltriethylammonium bromide

Warning

147125-100G 147125-25G
220036-418EA 73.08 USD
220036-418 220036-419
Benzyltriethylammonium bromide
Benzyltriethylammonium bromide
Formula: C₁₃H₂₂BrN
MW: 272.23 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5
EINECS: 225-986-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR