5-Methoxysalicylaldehyde ≥98%

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

146862-1G 146862-5G 146862-10G
220037-437EA 48.14 USD
220037-437 220036-410 220037-436
5-Methoxysalicylaldehyde ≥98%
5-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 51…52 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR