2-Indanone

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

146692-50G 146692-1G 146692-10G
220037-431EA 236.92 USD
220037-431 220037-430 220037-429
2-Indanone
2-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 220…225 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.0712 g/cm³ (67 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003792
CAS Number: 615-13-4
EINECS: 210-410-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR