Benzyltriethylammonium chloride

Warning

Synonyms: Benzyl triethylammonium chloride

146552-25G 146552-500G 146552-100G
220037-412EA 20.49 USD
220037-412 220037-413 220037-411
Benzyltriethylammonium chloride
Benzyltriethylammonium chloride
Formula: C₁₃H₂₂ClN
MW: 227.78 g/mol
Melting Pt: 190 - 192 °C
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR