Trimethyltin chloride

Danger

Synonyms: Chlorotrimethylstannane

146498-5G 146498-50G 146498-10G
220035-752EA 43.62 USD
220035-752 220035-751 220035-750
Trimethyltin chloride
Trimethyltin chloride
Formula: C₃H₉ClSn
MW: 199.27 g/mol
Boiling Pt: 96 °C (1013 hPa)
MDL Number: MFCD00000520
CAS Number: 1066-45-1
EINECS: 213-917-8
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR