1-Ethyl-2-pyrrolidone

Danger

Synonyms: 1-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, 1-Ethylpyrrolidin-2-one

146358-100G 146358-500G
220035-743EA 136.77 USD
220035-743 220035-744
1-Ethyl-2-pyrrolidone
1-Ethyl-2-pyrrolidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 97 °C (1013 hPa)
Density: 0.9979 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR