Bisacrylamide ≥99%

Warning

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

146072-100G 146072-500G
220037-382EA 46.25 USD
220037-382 220037-383
Bisacrylamide ≥99%
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Boiling Pt: 125 °C (3.33 hPa)
Melting Pt: 185 °C
Density: 1.235 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR