(-)-Quinine hydrochloride dihydrate

Danger

Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

145920-50G 145920-10G
220037-304EA 202.46 USD
220037-304 220037-303
(-)-Quinine hydrochloride dihydrate
(-)-Quinine hydrochloride dihydrate
Formula: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396.91 g/mol
Melting Pt: 158…160 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00151248
CAS Number: 6119-47-7
EINECS: 205-001-1

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR