(-)-Quinine

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

145904-10G 145904-50G
220037-299EA 91.75 USD
220037-299 220037-300
(-)-Quinine
(-)-Quinine
Formula: C₂₀H₂₄N₂O₂
MW: 324.42 g/mol
Melting Pt: 176 °C
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR