Allylamine ≥98%

Danger

Synonyms: 3-Aminopropene, 2-Propene-1-ylamine

145831-250ML 145831-1L
220037-294EA 100 USD
220037-294 220037-293
Allylamine ≥98%
Allylamine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 55…58 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.76 g/cm³ (20 °C)
Flash Pt: –29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008199
CAS Number: 107-11-9
EINECS: 203-463-9
UN: 2334
ADR: 6.1,I
Merck Index: 13,00284

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR