3-Methyl-2-cyclopenten-1-one ≥97%

Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone

145777-5G 145777-25G
220037-282EA 160.46 USD
220037-282 220037-281
3-Methyl-2-cyclopenten-1-one ≥97%
3-Methyl-2-cyclopenten-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 2 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001403
CAS Number: 2758-18-1
EINECS: 220-421-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR