DL-Tyrosine ≥99%

Synonyms: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH

145726-25G 145726-100G
220037-279EA 128.15 USD
220037-279 220037-278
DL-Tyrosine ≥99%
DL-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 285 °C
MDL Number: MFCD00063074
CAS Number: 556-03-6
EINECS: 209-113-1
Merck Index: 12,09970

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR