α-Bromo-m-toluonitrile ≥95%

Synonyms: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

145610-10G 145610-50G
220037-266EA 61.6 USD
220037-266 220037-267
α-Bromo-m-toluonitrile ≥95%
α-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 130 °C (4 -rr)
Melting Pt: 90 - 93 °C
MDL Number: MFCD00001809
CAS Number: 28188-41-2
EINECS: 248-890-1
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR