4-(Methylthio)benzoic acid ≥97%

145521-25G 145521-1G
220037-195EA 57.34 USD
220037-195 220037-194
4-(Methylthio)benzoic acid ≥97%
4-(Methylthio)benzoic acid
Formula: C₈H₈O₂S
MW: 168.22 g/mol
MDL Number: MFCD00002555
CAS Number: 13205-48-6
EINECS: 236-171-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR