2,3-Dimethyl-1,3-butadiene stabilized

Synonyms: 2,3-Dimethylbuta-1,3-diene

145491-50G 145491-10G
220037-189EA 329.54 USD
220037-189 220037-188
2,3-Dimethyl-1,3-butadiene stabilized
2,3-Dimethyl-1,3-butadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: -76 °C
Density: 0.7261 g/cm³ (20 °C)
MDL Number: MFCD00008595
CAS Number: 513-81-5
EINECS: 208-172-0
UN: 3295
ADR: 3,II
Merck Index: 13,03265

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR