4,6-Dichloropyrimidine

145378-10G 145378-50G
220036-767EA 94.45 USD
220036-767 220036-768
4,6-Dichloropyrimidine
4,6-Dichloropyrimidine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 64 - 66 °C
Density: 1.734 g/cm³ (20 °C)
MDL Number: MFCD00006109
CAS Number: 1193-21-1
EINECS: 214-770-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR