2,4-Dichlorotoluene ≥99%

Synonyms: 1,3-Dichloro-4-methylbenzene

145009-250G 145009-1KG
220037-088EA 61.06 USD
220037-088 220037-087
2,4-Dichlorotoluene ≥99%
2,4-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.2498 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000583
CAS Number: 95-73-8
EINECS: 202-445-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR