2,5-Dichlorobenzoic acid ≥97%

Warning

144940-250G 144940-50G
220037-082EA 334.92 USD
220037-082 220037-083
2,5-Dichlorobenzoic acid ≥97%
2,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 151 - 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3
EINECS: 200-065-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR