6-Amino-m-cresol ≥98%

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

144916-10G 144916-50G
220036-643EA 632.25 USD
220036-643 220036-644
6-Amino-m-cresol ≥98%
6-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 162 °C
MDL Number: MFCD00007693
CAS Number: 2835-98-5
EINECS: 220-620-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR