4-Amino-m-cresol ≥97%

Synonyms: 2-Amino-5-hydroxytoluene, 4-Amino-3-methylphenol, 1-Amino-4-hydroxy-2-methylbenzene, 4-Hydroxy-2-methylaniline

144894-5G 144894-25G
220036-641EA 76.57 USD
220036-641 220036-640
4-Amino-m-cresol ≥97%
4-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 177…179 °C
MDL Number: MFCD00007871
CAS Number: 2835-99-6
EINECS: 220-621-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR