3-(Methylthio)aniline ≥97%

Synonyms: 3-(Methylmercapto)aniline

144886-1G 144886-5G
220036-638EA 30.15 USD
220036-638 220036-639
3-(Methylthio)aniline ≥97%
3-(Methylthio)aniline
Formula: C₇H₉NS
MW: 139.22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Density: 1.1464 g/cm³ (20 °C)
MDL Number: MFCD00007793
CAS Number: 1783-81-9
EINECS: 217-232-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR