4-Phenylimidazole

Synonyms: 4-Phenyl-1H-imidazole

144754-10G 144754-5G
220036-629EA 103.71 USD
220036-629 220036-630
4-Phenylimidazole
4-Phenylimidazole
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 182…184 °C (1 mmHg)
Melting Pt: 144…146 °C
MDL Number: MFCD00005197
CAS Number: 670-95-1
EINECS: 211-580-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR