α-Bromo-o-toluonitrile ≥97%

Synonyms: 2-Cyanobenzyl bromide, 2-(Bromomethyl)benzonitrile, o-(Bromomethyl)benzonitrile

144711-5G 144711-100G 144711-25G
220036-628EA 50.83 USD
220036-628 220036-626 220036-627
α-Bromo-o-toluonitrile ≥97%
α-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 152…155 °C (15 mmHg)
Melting Pt: 72…74 °C
MDL Number: MFCD00001794
CAS Number: 22115-41-9
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR