4-Hydroxy-3-nitrobenzaldehyde ≥97%

Warning

144320-5G 144320-25G
220037-070EA 81.95 USD
220037-070 220037-069
4-Hydroxy-3-nitrobenzaldehyde ≥97%
4-Hydroxy-3-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007117
CAS Number: 3011-34-5
EINECS: 221-141-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR