2,4-Dichloro-6-methylpyrimidine

Danger

144185-100G 144185-25G
220037-055EA 165.85 USD
220037-055 220037-056
2,4-Dichloro-6-methylpyrimidine
2,4-Dichloro-6-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 47 °C
MDL Number: MFCD00006064
CAS Number: 5424-21-5
EINECS: 226-563-4
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR