3,5-Xylenol ≥99%

Danger

Synonyms: 5-Hydroxy-3-xylene, 3,5-Dimethylphenol, 5-Hydroxy-m-xylene, 1,3-Dimethyl-5-hydroxybenzene

144134-5G 144134-100G
220037-051EA 36.62 USD
220037-051 220037-050
3,5-Xylenol ≥99%
3,5-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 61 - 64 °C
Density: 0.968 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002307
CAS Number: 108-68-9
EINECS: 203-606-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR