4-Hydroxybenzaldehyde ≥98%

Warning

144088-50G 144088-250G
220037-046EA 37.58 USD
220037-046 220037-045
4-Hydroxybenzaldehyde ≥98%
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 116 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR