α-Bromo-p-toluonitrile ≥99%

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

144061-1G 144061-10G 144061-50G
220037-039EA 37.69 USD
220037-039 220037-038 220037-040
α-Bromo-p-toluonitrile ≥99%
α-Bromo-p-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 135…137 °C (0.6 mmHg)
Melting Pt: 115 °C
MDL Number: MFCD00001829
CAS Number: 17201-43-3
EINECS: 241-246-0
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR