Propargyl chloride ≥98%

Synonyms: 3-Chloropropyne, 3-Chloro-1-propyne

143995-25G 143995-5G
220037-023EA 103.17 USD
220037-023 220037-024
Propargyl chloride ≥98%
Propargyl chloride
Formula: C₃H₃Cl
MW: 74.51 g/mol
Boiling Pt: 58 °C (1013 hPa)
Melting Pt: –77 °C
Density: 1.0454 g/cm³ (5 °C)
MDL Number: MFCD00000980
CAS Number: 624-65-7
EINECS: 210-856-9
UN: 3286
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR