2,4,4-Trimethyl-2-pentene ≥99%

143820-5G 143820-25G
220037-013EA 104.25 USD
220037-013 220037-012
2,4,4-Trimethyl-2-pentene ≥99%
2,4,4-Trimethyl-2-pentene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 104 °C (1013 hPa)
Melting Pt: -106.3 °C
Density: 0.7223 g/cm³ (20 °C)
MDL Number: MFCD00008902
CAS Number: 107-40-4
EINECS: 203-488-5
UN: 2050
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR