Veratraldehyde ≥99%

Warning

Synonyms: 3,4-Dimethoxybenzaldehyde

143758-100G 143758-500G 143758-5G
220037-005EA 53.31 USD
220037-005 220037-006 220037-007
Veratraldehyde ≥99%
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 40 - 43 °C
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR