3-Hydroxy-p-anisaldehyde ≥99%

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

143685-100G 143685-25G
220036-999EA 102.42 USD
220036-999 220037-000
3-Hydroxy-p-anisaldehyde ≥99%
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR