2-(Bromomethyl)naphthalene

143677-5G 143677-100G 143677-25G
220036-998EA 69.57 USD
220036-998 220036-996 220036-997
2-(Bromomethyl)naphthalene
2-(Bromomethyl)naphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004123
CAS Number: 939-26-4
EINECS: 213-359-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR