2,5-Dimethyl-2,5-hexanediol ≥97%

Warning

Synonyms: 1,1,4,4-Tetramethyl-1,4-butanediol

143618-500G 143618-100G
220036-746EA 104.46 USD
220036-746 220036-745
2,5-Dimethyl-2,5-hexanediol ≥97%
2,5-Dimethyl-2,5-hexanediol
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 85 - 88 °C
Density: 0.898 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004473
CAS Number: 110-03-2
EINECS: 203-731-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR