2,5-Dimethoxycinnamic acid predominantly trans

Synonyms: 3-(2,5-Dimethoxyphenyl)acrylic acid, 2',5'-Dimethoxycinnamic acid, trans-2,5-Dimethoxycinnamic acid

143596-25G 143596-100G
220036-742EA 118.46 USD
220036-742 220036-741
2,5-Dimethoxycinnamic acid predominantly trans
2,5-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 150 °C
MDL Number: MFCD00004378
CAS Number: 10538-51-9
EINECS: 234-114-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR