5-Hydroxyindole-2-carboxylic acid

Synonyms: 5-Hydroxy-2-indolecarboxylic acid, 5-Hydroxy-1H-indole-2-carboxylic acid

143510-1G 143510-5G
220035-020EA 108.77 USD
220035-020 220036-738
5-Hydroxyindole-2-carboxylic acid
5-Hydroxyindole-2-carboxylic acid
Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00005615
CAS Number: 21598-06-1
EINECS: 244-468-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR