Diglycolic acid ≥98%

Synonyms: 2,2'-Oxydiacetic acid

143073-100G 143073-5G 143073-1KG
220036-933EA 107.05 USD
220036-933 220036-935 220036-934
Diglycolic acid ≥98%
Diglycolic acid
Formula: C₄H₆O₅
MW: 134.09 g/mol
MDL Number: MFCD00004309
CAS Number: 110-99-6
EINECS: 203-823-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR