Difluoroacetic acid ≥98%

Danger

142859-25G 142859-5G
220035-719EA 127.38 USD
220035-719 220035-720
Difluoroacetic acid ≥98%
Difluoroacetic acid
Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 134 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR