Chlorosulfonyl isocyanate ≥98%

142662-25G 142662-100G 142662-1KG
220036-869EA 70.22 USD
220036-869 220036-867 220036-868
Chlorosulfonyl isocyanate ≥98%
Chlorosulphonyl isocyanate
Formula: CClNO₃S
MW: 141.53 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.62 g/cm³ (20 °C)
MDL Number: MFCD00011608
CAS Number: 1189-71-5
EINECS: 214-715-2
UN: 3264
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR