2-Butyl-2-ethyl-1,3-propanediol ≥99%

Synonyms: 2-Ethyl-2-(hydroxymethyl)-1-hexanol, 3,3-Bis(hydroxymethyl)heptane, 2-Butyl-2-ethylpropane-1,3-diol

142476-500G 142476-100G
220036-862EA 166.92 USD
220036-862 220036-162
2-Butyl-2-ethyl-1,3-propanediol ≥99%
2-Butyl-2-ethyl-1,3-propanediol
Formula: C₉H₂₀O₂
MW: 160.26 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 39 °C
Density: 0.931 g/cm³ (50 °C)
MDL Number: MFCD00004697
CAS Number: 115-84-4
EINECS: 204-111-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR