4-tert-Butylpyridine

Warning

142379-25G 142379-100G
220036-154EA 121.15 USD
220036-154 220036-153
4-tert-Butylpyridine
4-tert-Butylpyridine
Formula: C₉H₁₃N
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –0.9 °C
Density: 0.9219 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006435
CAS Number: 3978-81-2
EINECS: 223-614-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR