Cycloheptene ≥97%

141720-25G 141720-5G
220036-513EA 384.46 USD
220036-513 220036-514
Cycloheptene ≥97%
Cycloheptene
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 114.6 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0.824 g/cm³ (20 °C)
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR