(±)-trans-1,2-Cyclohexanediol ≥98%

Synonyms: (±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol

141712-50G 141712-10G
220036-512EA 512.62 USD
220036-512 220036-511
(±)-trans-1,2-Cyclohexanediol ≥98%
(±)-trans-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 100…103 °C
Density: 1.147 g/cm³ (24 °C)
MDL Number: MFCD00063611
CAS Number: 1460-57-7
EINECS: 215-956-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR