2-Chloro-3-nitrobenzoic acid ≥99%

Warning

Synonyms: Mesalazine EP Impurity Q

141208-25G 141208-5G
220036-844EA 267.69 USD
220036-844 220036-845
2-Chloro-3-nitrobenzoic acid ≥99%
2-Chloro-3-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2
EINECS: 223-590-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR