Chloromethyl pivalate ≥97%

Synonyms: Chloromethyl trimethylacetate, Trimethylacetic acid chloromethyl ester, 2,2-Dimethylpropionic acid chloromethyl ester, Pivalic acid chloromethyl ester

141186-100G 141186-25G
220036-840EA 90.46 USD
220036-840 220036-841
Chloromethyl pivalate ≥97%
Chloromethyl pivalate
Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
Boiling Pt: 70 °C (1013 hPa)
Density: 1.0387 g/cm³ (20 °C)
MDL Number: MFCD00000884
CAS Number: 18997-19-8
EINECS: 242-735-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR