Isopropyl isocyanate ≥98%

141070-1G 141070-25G
220036-836EA 67.74 USD
220036-836 220036-837
Isopropyl isocyanate ≥98%
Isopropyl isocyanate
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 74 °C (1013 hPa)
Density: 0.8669 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002037
CAS Number: 1795-48-8
EINECS: 217-276-5
UN: 2483
ADR: 3,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR