3-Phenyl-1-propanol ≥98%

Synonyms: 3-Phenyl-1-Propanol

140856-250G 140856-50G
220036-820EA 69.68 USD
220036-820 220036-821
3-Phenyl-1-propanol ≥98%
3-Phenyl-1-propanol
Formula: (C₆H₅)CH₂CH₂CH₂OH
MW: 136.19 g/mol
Boiling Pt: 455 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4
EINECS: 204-587-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR