N,N-Dimethylformamide dimethyl acetal ≥94%, Technical Grade

Danger

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

140732-100ML 140732-25ML 140732-500ML
220036-802EA 90 USD
220036-802 220036-803 220036-804
N,N-Dimethylformamide dimethyl acetal ≥94%, Technical Grade
N,N-Dimethylformamide dimethyl acetal
Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 102 °C (1013 hPa)
Density: 0.893 g/cm³ (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00008482
CAS Number: 4637-24-5
EINECS: 225-063-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR