α-Chloro-4-nitrotoluene ≥99%

Synonyms: 4-Nitrobenzyl chloride, 4-Nitrobenzylchloride, α-Chloro-p-nitrotoluene

140112-25G 140112-100G
220035-908EA 70.86 USD
220035-908 220035-907
α-Chloro-4-nitrotoluene ≥99%
α-Chloro-4-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.5647 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR